Order Entry
Export
Orders LinkContactUsLinkComponent
Spray gun

Spray gun

Male connector for 1/2" O.Ø PVC tubing to 1/4" NPT inlet on gun
Catalog # ENTE215-98
Supplier:   ENTEGRIS
Spray gun
Catalog # ENTE215-98
Supplier:  ENTEGRIS
Spray gun
Spray gun

Male connector for 1/2" O.Ø PVC tubing to 1/4" NPT inlet on gun

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

High purity PFA and PTFE (all wetted parts).

  • Orifice: 4,8 mm
  • Connection: 1/4" or 1/2" FNPT
  • Pressure: max. 5516 mbar

Used in fume hoods and wet processing stations to help protect against corrosion.

Specifications
  • Description:
    Male connector for 1/2" O.Ø PVC tubing to 1/4" NPT inlet on gun
  • Seq:
    03