Orders LinkContactUsLinkComponent
Spray nozzles
Nozzle for spray gun
Catalog # ENTE215-66
Supplier:   ENTEGRIS
Spray nozzles
Catalog # ENTE215-66
Supplier:  ENTEGRIS
Spray nozzles
Nozzle for spray gun
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
Specifications
  • Description:
    Nozzle for spray gun