Mask carriers
Pocket spacing: 4,0 mm, pocket flat: various, 12 masks, PFA, Natural, 76x61x35 mm
Catalog # ENTEA41-01-0215
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Miscellaneous mask carriers.
Specifications
- For:12 masks
- Colour:Natural
- Description:Pocket spacing: 4,0 mm, pocket flat: various
- Material:PFA
- Seq:06
- W×D×H:76x61x35 mm