Orders LinkContactUsLinkComponent
Multiple wafer shippers
Multiple wafer shippers
Assembly, not cleaned, Wafershield™ high purity, Wafer thickness 150 - 675 µm, PP
Catalog # ENTEH9150-0037
Supplier:   ENTEGRIS
Multiple wafer shippers
Catalog # ENTEH9150-0037
Supplier:  ENTEGRIS
Multiple wafer shippers
Assembly, not cleaned, Wafershield™ high purity, Wafer thickness 150 - 675 µm, PP
Price
Inquire For Price
Log in for availability
1 * 12 items
Product Details & Documents
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.
  • Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
  • Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
  • Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
  • Three design options to accommodate varying wafer thickness
Specifications
  • For:
    Wafer thickness 150 - 675 µm
  • Description:
    Assembly, not cleaned, Wafershield™ high purity
  • Material:
    PP