Orders LinkContactUsLinkComponent
Gloves, HDPE
Gloves, HDPE
S, Transparent, 0,012 mm, 100/bag, 100 bags/box
100/box, 100 box/case
  HOIT110495
Gloves, HDPE
  HOIT110495
Gloves, HDPE
Gloves, HDPE
S, Transparent, 0,012 mm, 100/bag, 100 bags/box
100/box, 100 box/case
217,00 €
1 * 5.000 items
These disposable gloves are made from polyethene.
  • Glove size:
    S
  • Packed:
    100/bag, 100 bags/box
    100/box, 100 box/case
  • Colour:
    Transparent
  • Seq:
    01
  • Thickness:
    0,012 mm
  • Cat. No.:
    HOIT110495
  • Cuff thickness:
    0,012 mm
  • Disposable / reusable:
    Disposable