Order Entry
Export
Orders LinkContactUsLinkComponent
Nitrogen spray gun

Nitrogen spray gun

Nitrogen spray gun, natural
Catalog # ENTE421-42-11
Supplier:   ENTEGRIS
Nitrogen spray gun
Catalog # ENTE421-42-11
Supplier:  ENTEGRIS
Nitrogen spray gun
Nitrogen spray gun

Nitrogen spray gun, natural

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Long lasting and reliable PFA nitrogen spray guns with 0,5 µm PTFE membrane for use in high purity and corrosive chemical applications.

  • Integral filter enables air to be filtered before nitrogen is dispensed
  • Vented cap/nozzle meets OSHA safety specifications
  • PFA construction enables use in high purity and corrosive applications
  • Colored caps enable visual chemical differentiation at a glance

Nitrogen cap/nozzle prevents pressure build-up if outlet is blocked.

Orifice 4,8 mm
Max. pressure 75 psig (517 kPa)
Connection ¹/₄" FNPT

Accessories information: Repair kits are available.

Specifications
  • Description:
    Nitrogen spray gun, natural
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Repair tool to access front and rear diaphragm of gun
    Repair tool to access front and rear diaphragm of gun
    ENTE213-80
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap/nozzle, blue
    Replacement cap/nozzle, blue
    ENTE600082BL
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap/nozzle, natural
    Replacement cap/nozzle, natural
    ENTE600082
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap/nozzle, orange
    Replacement cap/nozzle, orange
    ENTE600082OR
    Log in for availability
    1 * 1 items