Bouffant caps, PP
White, 580 mm
Catalog # 113-8133
Supplier: HUBEI ORIENT INTL TRADING
Jump to:
Product Details & Documents
Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.
- Single elastic band
- Latex-free
- Soft and comfortable
Weight: 16 to 23 g/m²
Packaging: 100 pieces per polybag. 2000 pieces per carton.
Specifications
- Ø:580 mm
- Colour:White
- Seq:04
- Cat. No.:113-8133
- Garment application:Medical|Clinical



