ContactUsLinkComponent
Autoclav. goggle light blue #619042210 #619.04.22.10 + Comfortable, ergonomically designed full vision goggle + Can be worn over most prescription spectacles + Sophisticated indirect ventilation system + Panoramic field of vision white elastic strap 1 * 1 items
Catalog # UNVT619.04.22.10
Supplier:  UNIVET
undefined
Autoclav. goggle light blue #619042210 #619.04.22.10 + Comfortable, ergonomically designed full vision goggle + Can be worn over most prescription spectacles + Sophisticated indirect ventilation system + Panoramic field of vision white elastic strap 1 * 1 items
Catalog # UNVT619.04.22.10
Supplier:  UNIVET