Order Entry
Export
Orders LinkContactUsLinkComponent
 

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, left hinged, 600×366×480 mm
  KOTT307-00050
Wall-mounted cabinets
  KOTT307-00050
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, left hinged, 600×366×480 mm

 
 
   
1 * 1 items
 

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
 
  • Description:
    Wall-mounted cabinet, with one shelf
  • Seq:
    24
  • W×D×H:
    600×366×480 mm
  • Doors:
    1 door, left hinged