Order Entry
Export
Orders LinkContactUsLinkComponent
 

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 1 door, right hinged, 600×366×630 mm
Catalog # KOTT307-00058
Supplier:   KOETTERMANN
Wall-mounted cabinets
Catalog # KOTT307-00058
Supplier:  KOETTERMANN
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, lockable, 1 door, right hinged, 600×366×630 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Specifications
  • Description:
    Wall-mounted cabinet, with one shelf, lockable
  • Seq:
    29
  • W×D×H:
    600×366×630 mm
  • Doors:
    1 door, right hinged