Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 1 door, left hinged, 600×516×740 mm
Catalog # KOTT315-00070
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Description:Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
- Seq:14
- W×D×H:600×516×740 mm
- Doors:1 door, left hinged



