Mobile storage cabinets
Mobile storage cabinet, 1 door, right hinged, 600×516×590 mm
Catalog # KOTT315-00062
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Description:Mobile storage cabinet
- Seq:06
- W×D×H:600×516×590 mm
- Doors:1 door, right hinged