Order Entry
Export
Orders LinkContactUsLinkComponent
Mobile storage cabinets

Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door, left hinged, 600×516×740 mm
Catalog # KOTT315-00069
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00069
Supplier:  KOETTERMANN
Mobile storage cabinets

Mobile storage cabinet, with 1 drawer of 150 mm height, 1 door, left hinged, 600×516×740 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Epoxy powder coated steel cabinets

  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Description:
    Mobile storage cabinet, with 1 drawer of 150 mm height
  • Seq:
    13
  • W×D×H:
    600×516×740 mm
  • Doors:
    1 door, left hinged