VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/Conductivity/DO), Bench
Product Details & Documents
User friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. This meter can either measure pH/mV and conductivity or pH/mV and oxygen simultaneously. Complete with USB output for connection to a computer. GLP compliant.
- Large graphic LCD with continuous LED backlight shows the measured parameter and the temperature value simultaneously
- Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
- Automatic power shut-off: Adjustable from 10 min to 24 h
- Automatic buffer recognition of DIN and NIST buffers (1,68 / 4,00 / 6,86 / 9,18 / 12,54)
- Three additional technical buffers at 25 °C (4,00 / 7,00 / 10,00) in memory
- Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
- Low energy consumption. Choice of mains supply or 4× AA batteries for over 1000 operating hours
Ordering information: Conductivity probes for low conductivities are available on request. Please contact your local Avantor sales office for further information.
Delivery information: Ready to use MU 6100 L S1 kit (665-0309 ) includes bench multi-parameter meter, glass pH electrode pHenomenal 221 with built-in temperature sensor (1 m cable), conductivity probe CO 11 with built-in temperature sensor (1,5 m cable), articulated stand, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, short instructions and instruction manual (22 languages) on CD ROM.
The MU 6100 L S2 kit (665-0310) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal 221, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell CO 11, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, DO electrode OXY 11 (3 m cable).
Documents
665-0310
Images 360
Avantor Cell Culture Solutions.pdf
Product Literature
Specifications
- Description:Bench top pH/mV/conductivity/DO meter pHenomenal MU 6100 L S2 kit
- Connections:BNC for pH/banana for temperature/8-pin for conductivity and DO
- Compliance:IP 43/GLP
- DO concentration resolution:0,1%; 0,01 mg/L (depending on sensor)
- Automatic buffer recognition:VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
Technical buffers at 25 °C: 4,00/7,00/10,00 - DO accuracy:±0,5% of value (between 5...30 °C)
- Data logging capacity:5000 data sets
- mV resolution:0,1/1
- Resistivity range:1 Ω/cm - 999 MΩ/cm
- Conductivity accuracy:±0,5% of measured value
- DO calibration:Saturated air
- Temperature range:Auto: −5,0...+105,0 °C; manual: −25...+130 °C
- Salinity range:0,00 - 70 ppt
- mV range:±1200,0/±2500
- Resistivity accuracy:±0,5% of measured value
- pH range:−2,000...+19,999
- DO concentration:0,00 - 20,00 / 0,0 - 90,0 mg/L
- Temperature accuracy:±0,2 °C
- Cat. No.:665-0310
- Outputs:Waterproof USB connection (slave, bidirectional)
- TDS range:0 - 1999 mg/L / 2,00 - 19,99 g/L / 20,0 - 199,9 g/L
- mV accuracy:±0,3/±1
- Conductivity cell constant:0,001 - 25 cm⁻¹ (depending on probe)
- Conductivity resolution:0,1 µS/cm
- Power supply:Mains supply or 4× AA batteries
- Conductivity calibration:1 point
- Temperature compensation:Automatic/manual
- pH calibration:Up to 5 points
- Temperature resolution:0,1/1 °C
- TDS resolution:1 mg/L / 0,01 g/L / 0,1 g/L
- pH accuracy:±0,1/±0,01/±0,005
- pH resolution:0,1/0,01/0,001 (selectable)
- Data logging - log function:Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
- Model:MU 6100 L, S2 kit
- Conductivity range:0,001 µS/cm - 1000 mS/cm
- DO saturation:0,0 - 200,0; 0 - 600 % (depending on sensor)
- Display:Graphic LCD with LED backlight
Accessories
Product Information
- Catalog #Pack typePkAvailabilityPrice
- Potassium chloride 3 mol/L (3 N) in aqueous solution AVS TITRINORMPotassium chloride 3 mol/L (3 N) in aqueous solution AVS TITRINORM83605.180Plastic bottle100 mLLog in for availability1 * 100 mL - 32,30 €
Product Information
- Catalog #pH valuepH accuracypH buffer typeAvailabilityPrice
Product Information
- Catalog #Pack typePkAvailabilityPrice
- Electrode cleaning solution pepsin/hydrochloric acid for removal of proteinsElectrode cleaning solution pepsin/hydrochloric acid for removal of proteins83603.180Plastic bottle100 mLLog in for availability1 * 100 mL - 20,50 €
Product Information
- Catalog #pH valuepH accuracypH buffer typeAvailabilityPrice
Product Information
- Catalog #Pack typePkAvailabilityPrice
- Potassium chloride aqueous solution 3 mol/l (3 N) saturated with silver chloride aqueous solution AVS TITRINORM® for filling electrodesPotassium chloride aqueous solution 3 mol/l (3 N) saturated with silver chloride aqueous solution AVS TITRINORM® for filling electrodes83606.260Plastic bottle500 mLLog in for availability1 * 500 mL - 59,70 €
Product Information
- Catalog #AvailabilityPrice
- pHenomenal® storage/measuring vessel
Product Information
- Catalog #AvailabilityPrice
- pHenomenal® dissolved oxygen maintenance kit (incl. abrasive foil, electrolyte (50 ml), cleaning solution (30 ml), 3 exchange membrane caps)pHenomenal® dissolved oxygen maintenance kit (incl. abrasive foil, electrolyte (50 ml), cleaning solution (30 ml), 3 exchange membrane caps)664-0049Log in for availability1 * 1 items - 333,00 €
Product Information
- Catalog #AvailabilityPrice
- pHenomenal® electrode storage tube
Product Information
- Catalog #AvailabilityPrice
- pHenomenal electrode stand, holds up to 4 electrodes + 1 temperature sensorpHenomenal electrode stand, holds up to 4 electrodes + 1 temperature sensor662-1169Log in for availability1 * 1 items - 157,00 €
Product Information
- Catalog #AvailabilityPrice
- pHenomenal® field carrying case
Product Information
- Catalog #pH valuepH accuracypH buffer typeAvailabilityPrice



