Diallyl disulphide
Glass bottle for solids, 500 mg
CAYM10012582-500
: Diallyldisulphide, Allyl disulphide
- :2179-57-9
- :C₆H₁₀S₂
- :MFCD00008656
- :146.28 g/mol
- :198 °C (1013 hPa)
- :1.0237 g/cm³ (15 °C)
- :Refrigerator
- :218-548-6
- :2810
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:500 mg