Diallyl disulphide
Glass bottle for solids, 100 mg
Catalog # CAYM10012582-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Diallyldisulphide, Allyl disulphide
- CAS Number:2179-57-9
- Formula:C₆H₁₀S₂
- MDL Number:MFCD00008656
- Molecular Weight:146.28 g/mol
- Boiling Pt:198 °C (1013 hPa)
- Density:1.0237 g/cm³ (15 °C)
- Storage Temperature:Refrigerator
- EINECS:218-548-6
- UN:2810
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



