Order Entry
Export
Orders LinkContactUsLinkComponent
Diallyl disulphide

Diallyl disulphide

Glass bottle for solids, 1 g
Catalog # CAYM10012582-1
Irritant
Supplier:   Cayman Chemical
Diallyl disulphide
Catalog # CAYM10012582-1
Supplier:  Cayman Chemical
CAS Number:  2179-57-9
Diallyl disulphide

Glass bottle for solids, 1 g

CAS Number:  2179-57-9
Price
126,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Diallyldisulphide, Allyl disulphide
  • CAS Number:2179-57-9
  • Formula:C₆H₁₀S₂
  • MDL Number:MFCD00008656
  • Molecular Weight:146.28 g/mol
  • Boiling Pt:198 °C (1013 hPa)
  • Density:1.0237 g/cm³ (15 °C)
  • Storage Temperature:Refrigerator
  • EINECS:218-548-6
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications