Ionomycin calcium salt
Glass bottle for solids, 5 mg
Catalog # CAYM11932-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:56092-82-1
- Formula:C₄₁H₇₀CaO₉
- MDL Number:MFCD00151183
- Molecular Weight:747.1 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg



