Order Entry
Export
Orders LinkContactUsLinkComponent
 

Propidium Iodide

Glass bottle for solids, 100 mg
Catalog # CAYM14289-100
Irritant
Supplier:   Cayman Chemical
Propidium Iodide
Catalog # CAYM14289-100
Supplier:  Cayman Chemical
CAS Number:  25535-16-4
Propidium Iodide

Glass bottle for solids, 100 mg

CAS Number:  25535-16-4
Price
239,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • EINECS:247-081-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications