Order Entry
Export
Orders LinkContactUsLinkComponent
Heating tapes, HST

Heating tapes, HST

Heating tape HST with 5 m length, 250 W output
Catalog # HORS025206
Supplier:   HORST
Heating tapes, HST
Catalog # HORS025206
Supplier:  HORST
Heating tapes, HST

Heating tape HST with 5 m length, 250 W output

Price
214,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.

Specifications
  • Description:
    Heating tape HST with 5 m length, 250 W output
  • Seq:
    06