Orders LinkContactUsLinkComponent
Sterilin® Bottles, Water Sampling
350 ml, PET, Natural, Blue, Blue cap
Catalog # SLINWSC350
Supplier:   Sterilin
Sterilin® Bottles, Water Sampling
Catalog # SLINWSC350
Supplier:  Sterilin
Sterilin® Bottles, Water Sampling
Sterilin® Bottles, Water Sampling
350 ml, PET, Natural, Blue, Blue cap
Price
1 080,00 €
Log in for availability
1 * 144 items
Product Details & Documents
Blow moulded from flexible polystyrene.
  • Sterile by gamma irradiation
  • Dosed bottle (20 mg/L) is suitable for neutralising samples of low chlorinated water
  • Tamper evident cap for sample integrity
  • Lot number for complete traceability
  • Closure material: HDPE
Convenient way of sampling both chlorinated and non-chlorinated water for microbiological analysis.
Available manufactured using:
Flexible polystyrene
NEW, lightweight PET for improved handling
Polystyrene bottles feature color-coded labels to aid differentiation:
Blue: dosed
Green: undosed
Available dose levels:
20 mg/h: Suitable for neutralizing water samples with a low concentration of chlorine
120 mg/h: Suitable for neutralizing water samples with a high concentration of chlorine
Specifications
  • With:
    Blue cap
  • Capacity:
    350 ml
  • Colour:
    Natural
  • Material:
    PET
  • Seq:
    04
  • Cap colour:
    Blue