Orders LinkContactUsLinkComponent
Warning signs
Warning signs
Materials handling vehicles, PP, rigid, 200×200 mm
Catalog # BRDY252212
Supplier:   Brady
Warning signs
Catalog # BRDY252212
Supplier:  Brady
Warning signs
Warning signs
Materials handling vehicles, PP, rigid, 200×200 mm
Price
11,20 €
Log in for availability
1 * 1 items
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
  • Recommended by ISO 7010
  • Colour: Yellow/Black
  • Shape: Triangle
Specifications
  • Description:
    Materials handling vehicles
  • Dimensions:
    200×200 mm
  • Material:
    PP, rigid
  • Seq:
    25