ContactUsLinkComponent
Biotix 50ul 384 well clear full skirt PCR plate. 10 plates/pack, 5 packs/case. Ultra-thin , uniform thickness ensures consistent h eat transfer. polypropylene, RNase-, DNase-, and Endotoxin (pyrogen)-free. 1 * 10 items
Catalog # 732-2469
Supplier:  BIOTIX
Biotix 50ul 384 well clear full skirt PCR plate. 10 plates/pack, 5 packs/case. Ultra-thin , uniform thickness ensures consistent h eat transfer. polypropylene, RNase-, DNase-, and Endotoxin (pyrogen)-free. 1 * 10 items
Catalog # 732-2469
Supplier:  BIOTIX