Order Entry
Export
Orders LinkContactUsLinkComponent
Deep well microplates, 96-square well

Deep well microplates, 96-square well

Deep well microplate, 96-square well with V-bottom, Clear, -, 2,2 ml
Catalog # 732-2482
Supplier:   BIOTIX
Deep well microplates, 96-square well
Catalog # 732-2482
Supplier:  BIOTIX
Deep well microplates, 96-square well
Deep well microplates, 96-square well

Deep well microplate, 96-square well with V-bottom, Clear, -, 2,2 ml

Price
64,80 €
Log in for availability
1 * 5 items
Product Details & Documents

Medical grade virgin PP, non treated. These sturdy 96-deep well microplates conform to SBS standards for automated high throughput processing and compound storage.

  • Alphanumeric grid to help with sample identification; notched bottom right corner to ensure correct plate orientation
  • Conical shaped wells facilitate sample recovery
  • Each plate can withstand centrifugation up to 6000 RCF when supported by the appropriate microplate rotor
  • Excellent chemical resistance and temperature resistant from +121 to −80 °C
  • Stackable to save space in incubators and freezers
  • Tested free from DNase, RNase and endotoxin

Certifications: Conform to ANSI/SBS 1-2004 standards for automated workstations. All plates are BioReady™ certified for purity.

Documents

Specifications
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Deep well microplate, 96-square well with V-bottom
  • Seq:
    01
  • Well volume:
    2,2 ml