Order Entry
Export
Orders LinkContactUsLinkComponent
n-Dodecane

n-Dodecane

Glass bottle, 1
Catalog # METT51338012
Health
Supplier:   Mettler - Toledo
n-Dodecane
Catalog # METT51338012
Supplier:  Mettler - Toledo
CAS Number:  112-40-3
n-Dodecane
n-Dodecane

Glass bottle, 1

CAS Number:  112-40-3
Price
269,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:216 °C (1013 hPa)
  • Melting Pt:–9.6 °C
  • Density:0.7487 g/cm³ (20 °C)
  • Flash Pt:74 °C
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1

Specifications