0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Tissue Embedders
Accessories for Technovit® 9100
Accessories for Technovit® 9100
Basic solution, 5000 ml
HKUL66006735
:
HERAEUS KULZER
Accessories for Technovit® 9100
HKUL66006735
:
HERAEUS KULZER
Accessories for Technovit® 9100
Basic solution, 5000 ml
550,00 €
1 * 5 L
-
+
Description:
Basic solution, 5000 ml
Seq:
02