VWR® Q Path® Adhesion Slides
Adhesion slides, Q Path®, Green, ~1 mm
Supplier: LABONORD
Jump to:
Product Details & Documents
These slides can be used for the diagnostic testing of cells suspension and tissue samples.
- Store in a place with a stable temperature and low level humidity
Suitable for in vitro diagnostic (IVD) applications.
Certifications: EN 375:2001. ISO 8037/1.
Documents
Specifications
- Colour:Green
- Description:Adhesion slides, Q Path®
- Thickness:~1 mm