Orders LinkContactUsLinkComponent
VWR® Q Path® Adhesion Slides
VWR® Q Path® Adhesion Slides
White, ~1 mm
Catalog # 630-2087
Supplier:   LABONORD
VWR® Q Path® Adhesion Slides
Catalog # 630-2087
Supplier:  LABONORD
VWR® Q Path® Adhesion Slides
VWR® Q Path® Adhesion Slides
White, ~1 mm
Price
30,20 €
Log in for availability
1 * 72 items
Product Details & Documents
These slides can be used for the diagnostic testing of cells suspension and tissue samples.
  • Store in a place with a stable temperature and low level humidity
Suitable for in vitro diagnostic (IVD) applications.
Certifications: EN 375:2001. ISO 8037/1.
Specifications
  • Colour:
    White
  • Thickness:
    ~1 mm