Formamide ≥99.5%, Ultrapure
Plastic bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (1013 hPa)
- Melting Pt:2 °C
- Density:1.1334 g/cm³ (20 °C)
- Flash Pt:79 °C
- Storage Temperature:Refrigerator
- EINECS:200-842-0
Documents
Specifications
Specification Test Results
- Abs.@280 nm:≤0.05
- Conductivity:≤100 umhos
- Copper:<0.00001%
- DNase:None Detected
- Freezing Range:1 – 3 °C
- Iron:<0.0005%
- Lead:<0.00005%
- Moisture (KF):<0.75%
- Purity:>99.5%
- RNase:None
- Zinc:<0.00005%
Specifications
- Pack type:Plastic bottle
- Pk:500 mL



