Container for Hazardous Waste
Waste chute for container 736-0845, open and perforated on both sides, without "Biohazard" marking, 460×700×0,10 mm
Catalog # 736-0846
Supplier: HAMILTON ROBOTICS
Jump to:
Product Details & Documents
The box is used with waste chute 736-0847 and 736-0846.
- Container complete with yellow PE bag, thickness of foil 120 µm
- Two types of bags for container available
- Bags are highly resistant to tearing and puncturing
Specifications
- Description:Waste chute for container 736-0845, open and perforated on both sides, without "Biohazard" marking
- Seq:71
- W×D×H:460×700×0,10 mm



