Order Entry
Export
Orders LinkContactUsLinkComponent
Container for Hazardous Waste

Container for Hazardous Waste

Waste chute for container 736-0845, open and perforated on both sides, with "Biohazard" marking, 460×500×0,10 mm
Catalog # 736-0847
Supplier:   HAMILTON ROBOTICS
Container for Hazardous Waste
Catalog # 736-0847
Supplier:  HAMILTON ROBOTICS
Container for Hazardous Waste
Container for Hazardous Waste

Waste chute for container 736-0845, open and perforated on both sides, with "Biohazard" marking, 460×500×0,10 mm

Price
27,60 €
Log in for availability
1 * 10 items
Product Details & Documents

The box is used with waste chute 736-0847 and 736-0846.

  • Container complete with yellow PE bag, thickness of foil 120 µm
  • Two types of bags for container available
  • Bags are highly resistant to tearing and puncturing
Specifications
  • Description:
    Waste chute for container 736-0845, open and perforated on both sides, with "Biohazard" marking
  • Seq:
    72
  • W×D×H:
    460×500×0,10 mm