Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LICHROSORB 5 RP18 150 X 4.6MM 1 * 1 items
Catalog # VARIA0040150X046
Supplier:
VARIAN
Expand image
LICHROSORB 5 RP18 150 X 4.6MM 1 * 1 items
Catalog # VARIA0040150X046
Supplier:
VARIAN