Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AGILENT BIO SCX,NP1.7,4X10MM,SS GUARD 1 * 1 items
Catalog # HEWL5190-2422
Supplier:
Agilent
Expand image
AGILENT BIO SCX,NP1.7,4X10MM,SS GUARD 1 * 1 items
Catalog # HEWL5190-2422
Supplier:
Agilent