Order Entry
Export
Orders LinkContactUsLinkComponent
 

Formalin 10%, Q Path®, buffered

Plastic bottle, -, 1 L
Catalog # 11699455.
Health
Irritant
Supplier:   VWR Chemicals
Formalin 10%, Q Path®, buffered
Catalog # 11699455.
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formalin 10%, Q Path®, buffered
Formalin 10%, Q Path®, buffered

Plastic bottle, -, 1 L

CAS Number:  50-00-0
Price
20,60 €
Log in for availability
1 * 1 L
Product Details & Documents

Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.

Formaldehyde 4% is approximately equal to Formalin 10%.

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.013 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Appearance:
    Colourless liquid
  • Test formaldehyde:
    Passes test
  • Density (20/4):
    1,012 - 1,014
  • pH (20°C):
    6,8 - 7,2

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Pk Info:
    -

Specifications