Adjustable therm. with ground joint, AMA-Exact, -10+150:1C, enclosed scale, cap. blue reflecting, mercury filled, richter-top fini sh without button, lower part 300x8-8,5mm, upper part 250xc.10mm, suitable for government verification, with ground j 1 * 1 items
Adjustable therm. with ground joint, AMA-Exact, -10+150:1C, enclosed scale, cap. blue reflecting, mercury filled, richter-top fini sh without button, lower part 300x8-8,5mm, upper part 250xc.10mm, suitable for government verification, with ground j 1 * 1 items