Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Z-LINE FILTER F7 48MM 455X455MM 1 * 1 ST
Catalog # VOLZ11-000000-324
Supplier:
VOLZ LUFTFILTER
Expand image
Z-LINE FILTER F7 48MM 455X455MM 1 * 1 ST
Catalog # VOLZ11-000000-324
Supplier:
VOLZ LUFTFILTER