Pipette tips, ART™
ART™ 1000, 1000 µl, +, 88,9 mm, 8 racks, 96 each
Catalog # 613-2448
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
- Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
- Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
- Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
- Front latch closes and locks ensure safe and secure storage
- Space saving reload system is environmentally friendly, traceable and easy to use
Packaging: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.
Documents
Specifications
- Packed:8 racks, 96 each
- Sterile:+
- Volume:1000 µl
- Length:88,9 mm
- Seq:30
- Tip style:ART™ 1000
- Cat. No.:613-2448
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Empty hinged racks, medium, for 20 μl, 200 and 300 μl tipsEmpty hinged racks, medium, for 20 μl, 200 and 300 μl tips732-1170Log in for availability1 * 10 items - 43,90 €
Product Information
- Catalog #AvailabilityPrice
- Empty hinged racks, large, for 1000 μl tips
Product Information
- Catalog #AvailabilityPrice
- Empty hinged racks, small, for 10 μl and 20 μl tipsEmpty hinged racks, small, for 10 μl and 20 μl tips732-1171Log in for availability1 * 10 items - 43,40 €



