Orders LinkContactUsLinkComponent
Inoculating loops, Rotilabo®
Inoculating loop holder set, stainless steel, for inoculating loops with wire of Ø 0,6 - 1,0 mm, Assorted, 245 mm
  ROTHKL97.1
 :   Roth Carl
Inoculating loops, Rotilabo®
  ROTHKL97.1
 :  Roth Carl
Inoculating loops, Rotilabo®
Inoculating loop holder set, stainless steel, for inoculating loops with wire of Ø 0,6 - 1,0 mm, Assorted, 245 mm
75,20 €
1 * 4 items
Rotilabo® inoculating loops, made of platinum/iridium. Wire diameter 0,6 mm. Rotilabo® inoculating loop holder set made of stainless steel. Loops are fixed with union nut. For inoculating loops with wire diameter 0,6 to 1,0 mm. Set sorted by colour: blue, yellow, red, black. Length: 245 mm. Autoclavable at 121 °C.
 : Loops delivered without inoculating loop holder, please order separately.
  • Colour:
    Assorted
  • Description:
    Inoculating loop holder set, stainless steel, for inoculating loops with wire of Ø 0,6 - 1,0 mm
  • Length:
    245 mm
  • Seq:
    02