Level control systems with signalboxes, SymLine
Signalbox T1 with disc low level sensor, signal cable 3 meters, hook and loop fastener for disc sensor 2 meters, universal plug
Catalog # SCTV108157
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
- For all types of Safety Waste Caps and other electronic level controllers
- Touchless alarm-resetting fields for two containers
- 2 switch inputs and 2 switch outputs for peripheral units
LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface.
Specifications
- Description:Signalbox T1 with disc low level sensor, signal cable 3 meters, hook and loop fastener for disc sensor 2 meters, universal plug
- Seq:06
Accessories
Product Information
- Catalog #AvailabilityPrice
- Signal cable cable for signal transmission, 1,5 mSignal cable cable for signal transmission, 1,5 mSCTV108304Log in for availability1 * 1 items - 31,67 €
Product Information
- Catalog #AvailabilityPrice
- EU power supply
Product Information
- Catalog #AvailabilityPrice
- Dual lock (releasable pressure) closure for disc sensors, 20×20 mmDual lock (releasable pressure) closure for disc sensors, 20×20 mm554-2714Log in for availability1 * 10 items - 30,35 €
Product Information
- Catalog #AvailabilityPrice
- UK power supply
Product Information
- Catalog #AvailabilityPrice
- Velcro strip for disc sensor, 2 m
Product Information
- Catalog #AvailabilityPrice
- Extension cable for signal transmission, 5 m
Product Information
- Catalog #AvailabilityPrice
- Extension cable for signal transmission, 10 mExtension cable for signal transmission, 10 m590-0728Log in for availability1 * 1 items - 50,65 €
Product Information
- Catalog #AvailabilityPrice
- Cable for relay contact, 5 m
Product Information
- Catalog #AvailabilityPrice
- Disc sensor (alarm at empty state)
Product Information
- Catalog #AvailabilityPrice
- Disc sensor (alarm at full state)
Product Information
- Catalog #AvailabilityPrice
- Signal cable for electronic level control
Product Information
- Catalog #AvailabilityPrice
- Holder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HDHolder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HDSCTV106731Log in for availability1 * 1 items - 54,91 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp for level control, visual signal (flashing) when the level is reached, with magnetic holder for individual attachment, cable length of the lamp 3 m, power supply: 24 V DC EU/UK/US plugSignal lamp for level control, visual signal (flashing) when the level is reached, with magnetic holder for individual attachment, cable length of the lamp 3 m, power supply: 24 V DC EU/UK/US plugSCTV108334Log in for availability1 * 1 items - 218,23 €
Product Information
- Catalog #AvailabilityPrice
- Holder for disc sensor, GL 45
Product Information
- Catalog #AvailabilityPrice
- Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectorsSignal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectorsSCTV108334-TLog in for availability- 218,23 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp (full state) - EU/UK/US, for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment with disc sensor, visual alarm at full state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSignal lamp (full state) - EU/UK/US, for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment with disc sensor, visual alarm at full state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSCTV108335-TLog in for availability1 * 1 items - 404,99 €
Product Information
- Catalog #AvailabilityPrice
- Signal lamp (empty state) - EU/UK/US, for level control, visual and acoustic signal at empty state, with magnetic holder for individual attachment with disc sensor, visual alarm at empty state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSignal lamp (empty state) - EU/UK/US, for level control, visual and acoustic signal at empty state, with magnetic holder for individual attachment with disc sensor, visual alarm at empty state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plugSCTV108336-TLog in for availability1 * 1 items - 404,99 €