Colchicine ≥98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine
- CAS Number:64-86-8
- Formula:C₂₂H₂₅NO₆
- MDL Number:MFCD00078484
- Molecular Weight:399.44 g/mol
- Merck Index:14,2471
- Melting Pt:∼ 150 °C (dec.)
- Density:1.32 g/cm³ (20 °C)
- UN:1544
- ADR:6.1,I
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



