Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hydrogen generators, Precision Hydrogen series

Precision Hydrogen, 200cc, 200 ml/min, 380×540×406 mm
  564-0170
Hydrogen generators, Precision Hydrogen series
  564-0170
Hydrogen generators, Precision Hydrogen series
Hydrogen generators, Precision Hydrogen series

Precision Hydrogen, 200cc, 200 ml/min, 380×540×406 mm

 
8 060,00 €
   
1 * 1 items
 

Precision Hydrogen series hydrogen generators are suitable for carrier gas and flame gas at standard detection limits. Hydrogen purity: 99,9995%.

  • Proven PEM Technology generates hydrogen safely and reliably
  • Desiccant dryers ensure high level of purity
  • Automatic loading pump as standard
  • Maintenance limited to replacing de-ioniser cartridge and silica gel
  • Internal leak detection with automatic shutdown features

Delivery pressure: 0 to 6,9 bar

 
  • Flow rate:
    200 ml/min
  • Seq:
    02
  • W×D×H:
    380×540×406 mm
  • Type:
    Precision Hydrogen, 200cc
Accessories

 

  •  
    Description
     
     
  • PEAK05-7062
    Deioniser column
       
    1 * 1 items - 176,13 €