Orders LinkContactUsLinkComponent
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software renewal of 1 year software subscription
Catalog # 906-0097
Supplier:   VWR Collection
OpenLAB CDS EZChrom edition
Catalog # 906-0097
Supplier:  VWR Collection
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software renewal of 1 year software subscription
Price
771,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Scalable, compliant chromatography software platform with multi-instrument control. OpenLAB CDS EZChrom Edition is part of the Agilent software suite OpenLAB for analytical laboratories and enterprise. Its multi-instrument control capabilities, scalability and compliance are ideal to fulfil the requirements of today's chromatography laboratories, and to optimise their efficiency. Central administration functions have been arranged in the OpenLAB Shared Services/Control Panel module while keeping the existing user-interface and workflows for the EZChrom Elite™ chromatography data system. Many exciting functions are now available, from the Result Package mode and Result Review/Result Sequence mode to the new Intelligent Reporting mode. OpenLAB CDS EZChrom Edition can be easily complemented by further software modules belonging to the OpenLAB software suite.
  • Scalable from workstations to fully featured Client/Server systems
  • Configurable compliance
  • Multi-instrument control (many different LC and GC instrument control drivers available)
  • Intelligent reporting
  • Result Package mode
For further information and additional articles, please contact our specialists at Avantor or email [email protected].
Specifications
  • Description:
    OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software renewal of 1 year software subscription
  • Seq:
    05