Blank flanges, type KF
Blank flanges KF, DN 25, Aluminium
Catalog # ILMV701263
Supplier: Welch by Gardner Denver
Jump to:
Product Details & Documents
Blank flanges KF made of nickel-plated aluminium or stainless steel.
- Reliable
- Simple detachment for cleaning
Used for vacuum system.
Specifications
- Description:Blank flanges KF, DN 25
- Material:Aluminium