Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Probenröhren 12 ml, PS, N 95 Sn, steril 1 * 1.000 ST
Catalog # KABE050402
Supplier:
KABE LABORTECHNIK
Expand image
Probenröhren 12 ml, PS, N 95 Sn, steril 1 * 1.000 ST
Catalog # KABE050402
Supplier:
KABE LABORTECHNIK