Mirrors, Magoptic
Mirror, with focal length of +50 mm, 40 mm
Catalog # DIDAPOD608720
Supplier: DIDALAB
Jump to:
Product Details & Documents
Mirrors used for enabling classroom experiments.
Specifications
- Description:Mirror, with focal length of +50 mm
- Seq:04
- Ø:40 mm



