Mirrors, Magoptic
Mirror, with focal length of -100 mm, 80 mm
Catalog # DIDAPOD608480
Supplier: DIDALAB
Jump to:
Product Details & Documents
Mirrors used for enabling classroom experiments.
Specifications
- Description:Mirror, with focal length of -100 mm
- Seq:08
- Ø:80 mm



