Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Educational Materials
Physics Educational Materials
Optics Learning Activities
Screens for optical bench
Screens for optical bench
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm
DIDAPOD010006
:
DIDALAB
Screens for optical bench
DIDAPOD010006
:
DIDALAB
Screens for optical bench
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm
21,87 €
1 * 1 items
-
+
Description:
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm