Order Entry
Export
Orders LinkContactUsLinkComponent
He-Ne laser

He-Ne laser

He-Ne laser unpolarised, Max Power 1 mW, beam diameter 0,5 mm, 632,8 nm
Catalog # DIDAPOD013210
Supplier:   DIDALAB
He-Ne laser
Catalog # DIDAPOD013210
Supplier:  DIDALAB
He-Ne laser
He-Ne laser

He-Ne laser unpolarised, Max Power 1 mW, beam diameter 0,5 mm, 632,8 nm

Price
861,87 €
Log in for availability
1 * 1 items
Product Details & Documents

High quality professional gas laser, Class II

  • Max Power:<1 mW
  • Built-in tube power supply
  • Divergence <2 mrad for unpolarised laser

Delivery information: Supplied with rod Ø 10×130 mm for fitting to optical bench and mains power adapter

Specifications
  • Description:
    He-Ne laser unpolarised, Max Power 1 mW, beam diameter 0,5 mm
  • Wavelength:
    632,8 nm