Orders LinkContactUsLinkComponent
Laser protection spectacles
Laser protection spectacles
Red laser protection, 600/760 nm, Black, Blue
Catalog # DIDAPOD020500
Supplier:   DIDALAB
Laser protection spectacles
Catalog # DIDAPOD020500
Supplier:  DIDALAB
Laser protection spectacles
Laser protection spectacles
Red laser protection, 600/760 nm, Black, Blue
Price
83,13 €
Log in for availability
1 * 1 items
Product Details & Documents
Single lens safety glasses protect against red and blue laser.
  • For wavelengths 600/760 nm or 200/540 nm
  • Goggles are adjustable, one size fits all
Specifications
  • Type:
    Red laser protection, 600/760 nm
  • Colour:
    Black
  • Lenses:
    Blue