He-Ne laser
He-Ne laser polarised, Max Power 1 mW, 632,8 nm
Catalog # DIDAPOD013213
Supplier: DIDALAB
Jump to:
Product Details & Documents
High quality professional gas laser, Class II
- Max Power:<1 mW
- Built-in tube power supply
- Divergence <2 mrad for unpolarised laser
Delivery information: Supplied with rod Ø 10×130 mm for fitting to optical bench and mains power adapter
Specifications
- Description:He-Ne laser polarised, Max Power 1 mW
- Wavelength:632,8 nm



